Skip to Content
Merck
All Photos(2)

Key Documents

DWKWGW229P

Azlon Square Shoulder Unvented Wash Bottles

wide-neck, low-density polyethylene bottle, polypropylene closure (one of each color), capacity 250 mL

Synonym(s):

WGW229P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle
polypropylene closure (one of each color)
wide-neck

packaging

pkg of 1 ea

manufacturer/tradename

Azlon WGW229P

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Mixed pack of 5

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service