DWKBDA218P
Azlon Straight-sided Beakers
graduated, polypropylene, capacity 50 mL
Synonym(s):
BDA218P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
polypropylene
feature
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon
storage condition
room temperature
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Application
Ideal for general laboratory use and management of liquid samples
Features and Benefits
Straight sided stackable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service