Key Documents
DWK73770-20125
Kimble® Disposable Screw Thread Culture Tube, 25Ml
About This Item
UNSPSC Code:
41121706
NACRES:
NB.17
feature
Screw Thread Culture Tube
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73770-20125
capacity
25 mL
Related Categories
General description
These tubes are suitable for tissue culture work and general bacteriological use.
Application
Bacteriology, RIA, coagulation and other laboratory procedures
Features and Benefits
  • 73760 series and 90 series items that end in ‘F′ have flat bottoms which contribute to stability
  • 73770 series items are produced with a round bottom
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)