CLS4442125
Pyrex® long neck culture flask
capacity 125 mL
Synonym(s):
conical flasks, Corning® 4442
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
Recommended Products
material
Delong
Delong neck
borosilicate glass
flat bottom flask
sterility
non-sterile
feature
cap: no
closure type Closure cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 4442-125
capacity
125 mL
neck O.D.
25 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Neck is made from heavy-wall, rimless tubing and reduces the amount of splashed solution reaching the closure. Ideal for shaker cultures.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service