Skip to Content
Merck
All Photos(1)

Key Documents

713694

Sigma-Aldrich

(S)-(−)-2′-Amino-1,1′-binaphthalen-2-ol

97%

Synonym(s):

(S)-(−)-2-Amino-2′-hydroxy-1,1′-binaphthalene, (S)-NOBIN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO
CAS Number:
Molecular Weight:
285.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (HPLC)
97%

form

crystals

optical purity

enantiomeric excess: ≥98.0%

SMILES string

Nc1c(c4c(cc1)cccc4)c2c3c(ccc2O)cccc3

InChI

1S/C20H15NO/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12,22H,21H2

InChI key

HIXQCPGXQVQHJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-2′-Amino-1,1′-binaphthalen-2-ol or (S)-NOBIN, is a non-symmetrically substituted 1,1′-binaphthalene ligand. It can be used as a catalyst in the asymmetric synthesis of unnatural α-alkyl amino acids by phase-transfer catalysis (PTC). (S)-NOBIN can also be employed as starting material to prepare N-monoalkyl and N,N-dialkyl (S)-NOBIN analogs by reacting with aldehydes via reductive amination reaction. N-monoaryl derivatives are obtained from NOBIN via Hartwig−Buchwald arylation reaction.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2?-Amino-[1, 1?-binaphthalen]-2-ol
Kocovsky P
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service