Skip to Content
Merck
All Photos(2)

Key Documents

701513

Sigma-Aldrich

6-Methoxy-3-nitropyridine-2-acetonitrile

97%

Synonym(s):

(6-Methoxy-3-nitro-2-pyridyl)acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O3
CAS Number:
Molecular Weight:
193.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

117-122 °C

SMILES string

COc1ccc(c(CC#N)n1)[N+]([O-])=O

InChI

1S/C8H7N3O3/c1-14-8-3-2-7(11(12)13)6(10-8)4-5-9/h2-3H,4H2,1H3

InChI key

KOOPAOCWSWOMQI-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service