387789
Fluoride on polymer support
macroporous, 20-50 mesh, extent of labeling: 2.0-3.0 mmol/g loading
Synonym(s):
Amberlite™ IRA 900 F--form, Fluoride on Amberlyst® A-26
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H4CH2N(CH3)3F
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22
Recommended Products
form
macroporous
reaction suitability
reaction type: solution phase peptide synthesis
extent of labeling
2.0-3.0 mmol/g loading
particle size
20-50 mesh
Related Categories
Application
For a convenient synthesis of alkyl fluorides from alkyl halides or sulfonates.
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cainelli, G. et al.
Synthesis, 472-472 (1976)
Synthesis, 472-472 (1976)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service