1000MG7
Conjugated Polymer Nanoparticle (CPN)
fluorescence λem 1000 nm (IR-II), maleimide-thiol functionalized, 1 ea of 100 μg antibody kit
Synonym(s):
CPN™ 30-minute conjugation kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.51
Recommended Products
packaging
1 ea of 100 μg antibody kit
fluorescence
λem 1000 nm (IR-II)
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
CPNs are extremely bright and stable fluorophore nanoparticles which have been shown to give exceptional performance in a wide range of fluorescent analysis applications, such as lateral flow tests, fluorescent ELISA, cell tracking, immunofluorescence microscopy, flow cytometry and western blot. CPNs are resistant to photobleaching and a wide range of temperatures (-20°C to 120°C) and pH (2 to 12). They have been shown to be 1000x brighter than other fluorophores bringing exceptional levels of detection to assays and diagnostics.
Application
Thiol Antibody Conjugation kit allows covalent linkage reactions to be readily carried out without access to specialist chemistry capabilities. The kit is designed to be specific for CPN – antibody / protein linkage, has a rapid reaction time of 60 minutes in two steps, and comes in three sizes for the conjugation of 10mcg, 30mcg and 100mcg of antibodies. CPNs are are available in a range of emission wavelengths covering the visible and IR spectrum.
Legal Information
CPN is a trademark of Stream Bio Ltd
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service