Skip to Content
Merck
All Photos(1)

Key Documents

PHL85782

Violanthin

phyproof® Reference Substance

Synonym(s):

6-C-Glucosyl-8-C-rhamnosylapigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O=C1C=C(OC(C([C@@H]2O[C@H]([C@@H]([C@H]([C@H]2O)O)O)C)=C3O)=C1C(O)=C3[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)C5=CC=C(C=C5)O

InChI

1S/C27H30O14/c1-8-17(31)21(35)23(37)27(39-8)16-20(34)15(26-24(38)22(36)18(32)13(7-28)41-26)19(33)14-11(30)6-12(40-25(14)16)9-2-4-10(29)5-3-9/h2-6,8,13,17-18,21-24,26-29,31-38H,7H2,1H3/t8-,13+,17-,18+,21+,22-,23+,24+,26-,27-/m0/s1

InChI key

MVOUGOXRXQDXDC-RSPRXDBDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Violanthin is a compound that belongs to a class of organic compounds known as flavanoid 8-c-glycosides

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service