SYNF208450
Synthware™ three neck full jacket round bottom flask with vertical side necks
capacity 500 mL, center joint: ST/NS 29/42
Synonym(s):
Synthware F208450, 3-neck reaction flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
clear
Joint
center joint: ST/NS29/42
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F208450
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Flask, Round Bottom, Three Neck, Full Jacket, 500mL, 29/42 Joint
Application
kemetch; synthware; flask; round bottom; three neck
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service