Skip to Content
Merck
All Photos(2)

Documents

SAB5600281

Sigma-Aldrich

Anti-Mouse IgG Fab antibody, Goat monoclonal

recombinant, expressed in HEK 293 cells, clone RMG05, purified immunoglobulin

Synonym(s):

Fab region of Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RMG05, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

ELISA: 0.05-1 μg/mL
flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5—2 μg/mL
immunoprecipitation (IP): suitable (assay dependent concentration)
western blot: 0.1— 0.5 μg/mL

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

This antibody reacts to the Fab region of Mouse IgG, including IgG1, IgG2a, IgG2b, and IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.

Immunogen

Mouse IgG

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie-Xun Chen et al.
Oncology reports, 49(5) (2023-03-25)
The present study aimed to explore the potential roles of the methyltransferase‑like 3 (METTL3)‑mediated methylation of RAS related (RRAS) mRNA in the tumorigenesis and development of bladder cancer (BCa). For this purpose, the relative expression levels of METTL3 in BCa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service