Key Documents
HS1423A
Weigh Boat
Small, square black polystyrene, pk of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Pricing and availability is not currently available.
material
square black polystyrene
size
small
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1423A
L × W × H
(46 mm) × 46 mm × 8 mm
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer
Choose from one of the most recent versions:
Certificates of Analysis (COA)