Skip to Content
Merck
All Photos(1)

Key Documents

MCA-RH 7777

90021504, rat liver, Hepatocyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MCA-RH 7777, 90021504

biological source

rat liver

growth mode

Adherent

karyotype

Not specified

morphology

Hepatocyte

products

Alpha-foetoprotein

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Buffalo hepatoma

Cell Line Description

Derived from the Morris hepatoma 7777 (obtained from a male Buffalo strain rat). The original tumour was induced with N-2 fluorenylphthalamic acid. Particularly interesting for oncogeny studies since the cells produce α-fetoprotein (AFP).

Application

Tumourigenicity and oncogeny studies

Culture Medium

Swimm′s S-77 + 4mM Glutamine + 20% Horse Serum (HS) + 5% Foetal Bovine Serum (FBS) or EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures 1:2 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. NB: Cells are shed from the monolayer before confluency is achieved.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service