Skip to Content
Merck
All Photos(1)

Documents

PNT2 (SERUM FREE)

7042701, human prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PNT2 (SERUM FREE), 07042701

biological source

human prostate

growth mode

Adherent

karyotype

Not

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human prostate normal, serum-free

Cell Line Description

This cell line is a derivative of the normal human prostate cell line PNT2 (Sigma cat no. 95012613) adapted to grow in serum-free medium. PNT2 was established by immortalisation of normal adult prostatic epithelial cells by transfection with a plasmid containing SV40 genome with a defective replication origin. The primary culture was obtained from a prostate of a 33 year old male at post mortem. PNT2 cells contain the SV40 genome and express large T protein.

Culture Medium

Defined Keratinocyte-SFM (Gibco 10785) + Growth Supplement (Gibco 10784-015).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Use Trypsin Inhibitor to inactivate the trypsin. When freezing cells down use conditioned medium plus 10% DMSO.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service