Skip to Content
Merck
All Photos(2)

Key Documents

AV35262

Sigma-Aldrich

Anti-CLIC5 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Chloride intracellular channel 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
human, rabbit, rat, horse, mouse
citations:
3
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human, rabbit, rat, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLIC5(53405)

General description

CLIC5 is a chloride intracellular channel protein involved in hair cell stereocilia formation. It is reportedly stabilizes membrane-actin filament links at the base of stereocilia. It also has a role in the growth and differentiation of C2C12 myoblasts.
Rabbit Anti-CLIC5 antibody recognizes human, mouse, rat, pig, zebrafish, chicken, canine, and bovine CLIC5.

Immunogen

Synthetic peptide directed towards the middle region of human CLIC5

Application

Rabbit Anti-CLIC5 antibody is suitable for immunohistochemistry (4-8 μg/ml) and western blot (1.25 μg/ml) applications.

Biochem/physiol Actions

CLIon Channel5 specifically associates with the cytoskeleton of placenta microvilli. CLIon Channel-5A has a role as a chloride channel in vitro and binds to cortical actin cytoskeleton.

Sequence

Synthetic peptide located within the following region: HPPFLTFNGDVKTDVNKIEEFLEETLTPEKYPKLAAKHRESNTAGIDIFS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Carotenuto et al.
Nature communications, 12(1), 6738-6738 (2021-11-20)
FOLFIRINOX, a combination of chemotherapy drugs (Fluorouracil, Oxaliplatin, Irinotecan -FOI), provides the best clinical benefit in pancreatic ductal adenocarcinoma (PDAC) patients. In this study we explore the role of miRNAs (MIR) as modulators of chemosensitivity to identify potential biomarkers of
Fengna Li et al.
Cell biology international, 34(4), 379-384 (2010-01-09)
CLIC5 (chloride intracellular channel 5) is a CLIC (chloride intracellular channel) with various functions. Its high expression in skeletal muscle and association with actin-based cytoskeleton suggests that it may play an important role in muscle tissue. This study was conducted
Felipe T Salles et al.
Cytoskeleton (Hoboken, N.J.), 71(1), 61-78 (2013-11-29)
Chloride intracellular channel 5 protein (CLIC5) was originally isolated from microvilli in complex with actin binding proteins including ezrin, a member of the Ezrin-Radixin-Moesin (ERM) family of membrane-cytoskeletal linkers. CLIC5 concentrates at the base of hair cell stereocilia and is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service