Skip to Content
Merck
All Photos(1)

Key Documents

BCR093R

1-Methylbenz[a]anthracene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14
CAS Number:
Molecular Weight:
242.31
Beilstein:
2098160
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

Cc1cccc2ccc3cc4ccccc4cc3c12

InChI

1S/C19H14/c1-13-5-4-8-14-9-10-17-11-15-6-2-3-7-16(15)12-18(17)19(13)14/h2-12H,1H3

InChI key

MWMBCXXTFFHALK-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR093R

Legal Information

BCR is a registered trademark of European Commission

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Glatt et al.
Mutagenesis, 9(2), 83-92 (1994-03-01)
The K-region oxides and imines of benz[a]anthracene, 1-methylbenz[a]anthracene, 7-methylbenz[a]anthracene, 7-ethylbenz[a]anthracene and 7,12-dimethylbenz[a]anthracene were synthesized and characterized (melting point, 1H-NMR and electron impact mass spectra, elemental analysis, IR spectroscopy). All 10 compounds showed high mutagenic activity in Salmonella typhimurium (reversion of
[Induction of tumors by chemical carcinogens. Experimental study].
J A Sánchez Cousteau et al.
Revista clinica espanola, 161(2), 95-97 (1981-04-30)
William B Studabaker et al.
Journal of chromatography. A, 1492, 106-116 (2017-03-08)
Development of the Athabasca Oil Sands Region in northeastern Alberta, Canada has contributed polycyclic aromatic hydrocarbons (PAHs) and polycyclic aromatic compounds (PACs), which include alkyl PAHs and dibenzothiophenes, to the regional environment. A new analytical method was developed for quantification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service