Skip to Content
Merck
All Photos(1)

Documents

MABE1886

Sigma-Aldrich

Anti-BMAL1 (ARNTL) Antibody, clone B1BH2

clone B1BH2, from mouse

Synonym(s):

Aryl hydrocarbon receptor nuclear translocator-like protein 1, BMAL1b′, Brain and muscle ARNT-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B1BH2, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), rat (based on 100% sequence homology), chicken (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

chicken ... Arntl(374115)
human ... ARNTL(406)
mouse ... Arntl(11865)
rat ... Arntl(29657)

General description

Aryl hydrocarbon receptor nuclear translocator-like protein 1 (UniProt: Q9I8T7; also known as BMAL1b′, Brain and muscle ARNT-like 1, cBMAL1) is encoded by the ARNTL (also known as BMAL1) gene (Gene ID: 374115) in chicken. BMAL1 is expressed in pineal gland and retina and displays maximum levels between ZT13 and ZT16 during light/dark cycle. BMAL1 is a transcriptional activator that forms a core component of the circadian clock. These components include ARNTL/BMAL1, CLOCK, NPAS2, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1, and CRY2. CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 form the positive limb of the feedback loop. They act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes that are involved in key metabolic processes. PER1/2/3 and CRY1/2, act as transcriptional repressors and form the negative limb of the feedback loop. They interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity. BMLA1 is highly expressed in the adult brain, skeletal muscle, and heart. It shuttles between the nucleus and the cytoplasm and this nucleocytoplasmic shuttling is essential for the nuclear accumulation of CLOCK, target gene transcription, and the degradation of the CLOCK-ARNTL/BMAL1 heterodimer. BMLA1 is reported to be phosphorylated upon dimerization with CLOCK and this phosphorylation enhances its transcriptional activity and reduces the stability of the CLOCK-ARNTL/BMAL1 heterodimer by promoting its degradation. BMAL1 null-mice are reported to have reproductive problems, reduced locomotor activity, and premature aging.

Specificity

Clone B1BH2 specifically detects Brain and muscle ARNT-like 1 (BMAL1). It targets an epitope within 56 amino acids from the N-terminal region.

Immunogen

MBP-conjugated recombinant fragment corresponding to 56 amino acids from the N-terminal region of chicken BMAL1.

Application

Anti-BMAL1 (ARNTL), clone B1BH2, Cat. No. MABE1886, is a highly specific mouse monoclonal antibody that targets Brain and muscle ARNT-like 1 (BMAL1) and has been tested for use in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
Chromatin Immunoprecipitation Analysis (ChIP): A representative lot detected BMAL1 (ARNTL) in Chromatin Immunoprecipitation applications (Yoshitane, H., et. al. (2014). Mol Cell Biol. 34(10):1776-87).

Western Blotting Analysis: A representative lot detected BMAL1 (ARNTL) in Western Blotting applications (Yoshitane, H., et. al. (2009). Mol Cell Biol. 29(13):3675-86; Yoshitane, H., et. al. (2012). EMBO Rep. 13(5):455-61).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected BMAL1 (ARNTL) in NIH/3T3 cell lysate.

Target description

~75 kDa observed; 69.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service