Skip to Content
Merck
All Photos(1)

Key Documents

8.16113

Sigma-Aldrich

1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione

(stabilised) for synthesis

Synonym(s):

1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric acid triallyl ester, Triallyl isocyanurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
MDL number:
UNSPSC Code:
12352115
EC Index Number:
213-834-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

3.5 hPa ( 143 °C)

Quality Level

Assay

≥98.0% (GC)

potency

700 mg/kg LD50, oral (Rat)
2480 mg/kg LD50, skin (Rat)

kinematic viscosity

78.2 cSt(30 °C)

bp

150 °C/5.3 hPa

mp

25 °C

transition temp

flash point 160 °C

solubility

3.7 g/L

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(C(=O)N(C(=O)N(C1=O)CC=C)CC=C)CC=C

InChI

1S/C12H15N3O3/c1-4-7-13-10(16)14(8-5-2)12(18)15(9-6-3)11(13)17/h4-6H,1-3,7-9H2

InChI key

KOMNUTZXSVSERR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.158 - 1.162
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.1 °F - closed cup

Flash Point(C)

159.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service