Key Documents
Z562122
Platinum crucible lid
fits 50 mL crucible
Synonym(s):
crucibles
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Birmingham metals
weight
10.0 g
General description
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Choose from one of the most recent versions:
Certificates of Analysis (COA)