Key Documents
SYNF614125
Synthware pressure equalizing addition funnel with metering stopcock
125 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F614125, dropping funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F614125
capacity
125 mL
125 mL
Related Categories
General description
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)