BR472800
BRAND® glass staining trough, Coplin pattern
soda-lime glass
Synonym(s):
Microscopy, Staining, Staining Accessories
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
Recommended Products
material
soda-lime glass
manufacturer/tradename
BRAND 472800
L × W
76 mm × 26 mm , holds 10 slides
Related Categories
General description
The BRAND® Glass Staining Trough in the Coplin pattern is specifically designed for the efficient staining of up to 10 slides, making it an essential tool for laboratory procedures. Constructed from durable soda-lime glass, this trough has dimensions of 76 x 26 mm, providing a compact yet effective solution for slide handling. Its Coplin design ensures optimal spacing for slides, facilitating uniform staining and easy access. This reliable and practical staining trough enhances the efficiency of laboratory workflows, making it a valuable addition to any histology or microscopy setup.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Doug K Tischer et al.
eLife, 8 (2019-04-06)
T cells are thought to discriminate self from foreign peptides by converting small differences in ligand binding half-life into large changes in cell signaling. Such a kinetic proofreading model has been difficult to test directly, as existing methods of altering