Skip to Content
Merck
All Photos(1)

Key Documents

933007

Sigma-Aldrich

Wafer Tweezers

Style 43LB-8 Peek

Synonym(s):

Wafer Tweezers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

Wafer Tweezers Style 43LB-8 Peek, made from Polyether ether ketone (PEEK), an Electrostatic discharge (ESD) safe soft, conductive fiber, with heat resistance up to 250 °C. Clean-room compatible. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers. They have a smooth polished non-glare surface with a satin finish. The tweezers have stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service