Skip to Content
Merck
All Photos(1)

Key Documents

512370

Sigma-Aldrich

6-Amino-2-naphthoic acid

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6CO2H
CAS Number:
Molecular Weight:
187.19
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

reaction suitability

reaction type: solution phase peptide synthesis

impurities

3% 5-amino-2-naphthoic acid
5% 6-hydroxy-2-naphthoic acid

mp

206-209 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc2cc(ccc2c1)C(O)=O

InChI

1S/C11H9NO2/c12-10-4-3-7-5-9(11(13)14)2-1-8(7)6-10/h1-6H,12H2,(H,13,14)

InChI key

NZTPZUIIYNYZKT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven J Cartwright
Homeopathy : the journal of the Faculty of Homeopathy, 107(1), 19-31 (2018-03-13)
The use of solvatochromic dyes to investigate homeopathic potencies holds out the promise of understanding the nature of serially succussed and diluted solutions at a fundamental physicochemical level. Recent studies have shown that a range of different dyes interact with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service