コンテンツへスキップ
Merck
  • Diosgenin, an Activator of 1,25D3-MARRS Receptor/ERp57, Attenuates the Effects of TNF-α by Causing ADAM10-Dependent Ectodomain Shedding of TNF Receptor 1.
Diosgenin, an Activator of 1,25D3-MARRS Receptor/ERp57, Attenuates the Effects of TNF-α by Causing ADAM10-Dependent Ectodomain Shedding of TNF Receptor 1.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-27)
Won Seok Yang, Soo Young Moon, Mee Jeong Lee, Eun Kyoung Lee, Su-Kil Park
要旨
We investigated how diosgenin, a steroidal sapogenin, has anti-tumor necrosis factor-α (TNF-α) effects in human aortic endothelial cells (HAECs). Tumor necrosis factor receptor 1 (TNFR1) was assessed by Western blot analysis. Intracellular Ca2+ was measured using Fluo-4 AM. Immunofluorescence staining was performed for a disintegrin and metalloprotease 10 (ADAM10). Diosgenin (1 ∼ 100 nM) induced ectodomain shedding of TNFR1 within 30 min and attenuated TNF-α-induced intercellular adhesion molecule-1 (ICAM-1) expression. Upon treatment with diosgenin, extracellular Ca2+ entered into the cells via L-type calcium channels, whereas diosgenin-induced ectodomain shedding of TNFR1 was almost completely inhibited by BAPTA-AM (intracellular Ca2+ chelator), verapamil (L-type calcium channel antagonist) and the absence of extracellular Ca2+. Diosgenin caused translocation of ADAM10 to the cell surface, which was mediated by extracellular Ca2+ influx. Depletion of ADAM10 prevented diosgenin-induced ectodomain shedding of TNFR1 and abolished the inhibitory effect of diosgenin on TNF-α-induced ICAM-1 expression. Diosgenin did not induce extracellular Ca2+ influx and ectodomain shedding of TNFR1 in cells depleted of 1,25D3-membrane associated rapid response steroid-binding receptor (1,25D3-MARRS receptor/ERp57). Diosgenin elicits L-type calcium channel-mediated extracellular Ca2+ influx, and thereby induces ADAM10-mediated ectodomain shedding of TNFR1. This effect of diosgenin was exerted through 1,25D3-MARRS receptor/ERp57.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジオスゲニン, ≥93%