コンテンツへスキップ
Merck
  • The Mucosal Antibacterial Response Profile and Fecal Microbiota Composition Are Linked to the Disease Course in Patients with Newly Diagnosed Ulcerative Colitis.
The Mucosal Antibacterial Response Profile and Fecal Microbiota Composition Are Linked to the Disease Course in Patients with Newly Diagnosed Ulcerative Colitis.
Inflammatory bowel diseases (2017-04-27)
Maria K Magnusson, Hans Strid, Stefan Isaksson, Magnus Simrén, Lena Öhman
要旨
The clinical disease course of ulcerative colitis (UC) varies substantially between individuals and can currently not be reliably predicted. The gut microbiota and the host's immune defense are key players for gut homeostasis and may be linked to disease outcome. The aim of this study was to determine fecal microbiota composition and mucosal antibacterial response profile in untreated patients with newly diagnosed UC and the impact of these factors on disease course. Stool samples and intestinal biopsies were obtained from therapy-naive newly diagnosed patients with UC. Patients were defined to have mild or moderate/severe disease course assessed by disease activity during the 3 years follow-up. Fecal microbiota was analyzed by the GA-map Dysbiosis test (n = 18), and gene expression in intestinal biopsies was analyzed by RT Profiler polymerase chain reaction array (n = 13) and real-time polymerase chain reaction (n = 44). At the time of diagnosis of UC, the fecal microbiota composition discriminated between patients with mild versus moderate/severe disease course. Also, the mucosal antibacterial gene expression response profile differed between patients with mild versus moderate/severe disease course with bactericidal/permeability-increasing protein (BPI) being most important for the discrimination. Mucosal bactericidal/permeability-increasing protein gene expression at diagnosis was higher in patients with mild versus moderate/severe disease course when confirmed in a larger patient cohort (P = 0.0004, n = 44) and was a good predictor for the number of flares during the 3 years follow-up (R = 0.395, P < 0.0001). In patients with newly diagnosed UC, fecal microbiota composition and mucosal antibacterial response profile, especially bactericidal/permeability-increasing protein, are linked to disease course.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
7-アミノアクチノマイシンD, ~97% (HPLC), powder