コンテンツへスキップ
Merck
  • Mechanism of Ion-Pair Strategy in Modulating Skin Permeability of Zaltoprofen: Insight from Molecular-Level Resolution Based on Molecular Modeling and Confocal Laser Scanning Microscopy.
Mechanism of Ion-Pair Strategy in Modulating Skin Permeability of Zaltoprofen: Insight from Molecular-Level Resolution Based on Molecular Modeling and Confocal Laser Scanning Microscopy.
Journal of pharmaceutical sciences (2015-09-10)
Hongxia Cui, Peng Quan, Hanqing Zhao, Xue Wen, Wenting Song, Yi Xiao, Yongshan Zhao, Liang Fang
要旨
The study was carried out to investigate the mechanism of the ion-pair strategy in modulating zaltoprofen (ZAL) skin permeability. Seven organic amines were chosen as counter ions and the formation of ion pairs was confirmed by Fourier transform infrared and proton nuclear magnetic resonance spectroscopy. In vitro permeation studies showed that the permeation of ZAL was significantly promoted with alkylamines and cycloalkanolamines, while significantly retarded with alkanolamines. The results were further visualized by confocal laser scanning microscopy studies. Investigations from the standpoint of solubility, partition, and diffusion indicated that the modulation mechanism of ion pairs was mainly attributed to an alteration of ZAL diffusion in the skin, which was accomplished by masking the carboxyl of ZAL and adding the hydrogen-bonding groups of counter ions. A negative correlation was observed between the enhancement ratio of ion pairs and the polar surface area of counter ions. Molecular modeling revealed that ion pairs acted with the nature of different affinity to ceramide, which would be responsible for the diffusion retardation. The study indicated that the maximum effect of ion pairs tended to be achieved with counter ions of lower hydrogen-bonding potential. Besides, ZAL-triethylamine ion pair might be a promising strategy for transdermal delivery of ZAL.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
1-(2-ヒドロキシエチル)ピペリジン, ReagentPlus®, 99%