コンテンツへスキップ
Merck
  • Highly enantioselective synthesis of nitrocyclopropanes via organocatalytic conjugate addition of bromomalonate to alpha,beta-unsaturated nitroalkenes.
Highly enantioselective synthesis of nitrocyclopropanes via organocatalytic conjugate addition of bromomalonate to alpha,beta-unsaturated nitroalkenes.
Organic letters (2009-03-06)
Yi-Ning Xuan, Shao-Zhen Nie, Li-Ting Dong, Jun-Min Zhang, Ming Yan
要旨
Highly enantioselective synthesis of nitrocyclopropanes was achieved via the organocatalytic conjugate addition of dimethyl bromomalonate to nitroalkenes and the consequent intramolecular cyclopropanation. 6'-Demethyl quinine was found to be the efficient catalyst. Excellent enantioselectivities, diastereoselectivities, and good yields were obtained for a variety of aryl or heteroaryl nitroethylenes.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ブロモマロン酸ジメチル, technical grade, 90%