コンテンツへスキップ
Merck
  • Determination of alpha-amanitin and beta-amanitin in human biological fluids by high-performance liquid chromatography.
Determination of alpha-amanitin and beta-amanitin in human biological fluids by high-performance liquid chromatography.
Analytical biochemistry (1985-08-15)
F Jehl, C Gallion, P Birckel, A Jaeger, F Flesch, R Minck
要旨
A high-performance liquid chromatographic assay of alpha-amanitin and beta-amanitin in human serum, urine, or stomach washings is described. Sample preparation involves a chemical step with deproteinization and organic solvent treatment, and a selective cleanup and concentration step on reversed-phase prepacked cartridges. Separations are performed on a reversed-phase analytical column under isocratic conditions with uv detection at 280 nm. The method allows the quantitation of alpha- and beta-amanitin separately with a detection limit of 10 ng/ml for both toxins.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
β-アマニチン from Amanita phalloides, ≥85% (HPLC)