コンテンツへスキップ
Merck
Direct N(2)H(4)/H(2)O(2) fuel cells powered by nanoporous gold leaves.
Scientific reports (2012-12-12)
Xiuling Yan, Fanhui Meng, Yun Xie, Jianguo Liu, Yi Ding
要旨
Dealloyed nanoporous gold leaves (NPGLs) are found to exhibit high electrocatalytic properties toward both hydrazine (N(2)H(4)) oxidation and hydrogen peroxide (H(2)O(2)) reduction. This observation allows the implementation of a direct hydrazine-hydrogen peroxide fuel cell (DHHPFC) based on these novel porous membrane catalysts. The effects of fuel and oxidizer flow rate, concentration and cell temperature on the performance of DHHPFC are systematically investigated. With a loading of ~0.1 mg cm(-2) Au on each side, an open circuit voltage (OCV) of 1.2 V is obtained at 80°C with a maximum power density 195 mW cm(-2), which is 22 times higher than that of commercial Pt/C electrocatalyst at the same noble metal loading. NPGLs thus hold great potential as effective and stable electrocatalysts for DHHPFCs.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ヒドラジン 一水和物, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
ヒドラジン 溶液, 35 wt. % in H2O
Sigma-Aldrich
ヒドラジン 硫酸塩, ACS reagent, ≥99.0%
Sigma-Aldrich
ヒドラジン 溶液, 1.0 M in THF
Sigma-Aldrich
ヒドラジン 溶液, 1.0 M in ethanol
Sigma-Aldrich
ヒドラジン 硫酸塩, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
ヒドラジン 二塩酸塩, ≥98%
Sigma-Aldrich
Hydrazinium dichloride, SAJ special grade, ≥98.0%
Sigma-Aldrich
ヒドラジン 溶液, 1 M in acetonitrile
Sigma-Aldrich
ヒドラジン ヘミ硫酸塩, ≥98%
Sigma-Aldrich
ヒドラジン 一水和物, SAJ first grade, ≥98.0%
Sigma-Aldrich
Hydrazinium sulfate, SAJ first grade, ≥97.0%