コンテンツへスキップ
Merck
  • Knockdown of TRIM5α or TRIM11 increases lentiviral vector transduction efficiency of human Muller cells.
Knockdown of TRIM5α or TRIM11 increases lentiviral vector transduction efficiency of human Muller cells.
Experimental eye research (2021-01-14)
Monica M Sauter, Curtis R Brandt
要旨
The goal of this study was to determine the expression and distribution of the host restriction factors (RFs) TRIM5α and TRIM11 in non-human primate (NHP) neural retina tissue and the human Muller cell line MIO-M1. In addition, experiments were performed to determine the effect of TRIM5α and TRIM11 knockdown on FIVGFP transduction of MIO-M1 cells with the goal of devising strategies to increase the efficiency of lentiviral (LV) gene delivery. Immunofluorescence (IF) studies indicated that TRIM5α and TRIM11 were localized predominantly in nuclei within the outer nuclear layer (ONL) and inner nuclear layer (INL) of NHP retina tissue. Double label IF indicated that TRIM5α and TRIM11 were localized to some of the retinal Muller cell nuclei. MIO-M1 cells expressed TRIM5α predominantly in the nucleus and TRIM11 primarily in the cytosol. FIVGFP transduction efficiency was significantly increased, at 4 and 7 days post transduction, in TRIM5α and TRIM11 knockdown clones (KD) compared to WT MIO-M1 cells. In addition, pretreatment with the proteasome inhibitor MG132 increased the transduction efficiency of FIVGFP in WT MIO-M1 cells. The nuclear translocation of NF-κB (p65), at 72 h post FIVGFP transduction, was enhanced in TRIM5α and TRIM11 KD clones. The expression of TRIM5α and TRIM11 in macaque neural retina tissue and MIO-M1 cells indicate the presence of these RFs in NHP retina and human Muller cells. Our data indicate that even partial knockdown of TRIM5α or TRIM11, or a short proteasome inhibitor pretreatment, can increase the transduction efficiency of a LV vector.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
プロテアーゼインヒビターカクテル, for use with mammalian cell and tissue extracts, DMSO solution
カタログ番号
容量
在庫状況
単価
数量
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
カタログ番号
容量
在庫状況
単価
数量
Sigma-Aldrich
抗FLAGタグ ウサギ宿主抗体, affinity isolated antibody
カタログ番号
容量
在庫状況
単価
数量
Sigma-Aldrich
Phosphatase Inhibitor Cocktail Set IV, Lyophilized, The Phosphatase Inhibitor Cocktail Set IV, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
カタログ番号
容量
在庫状況
単価
数量