コンテンツへスキップ
Merck
  • Self-Suspended Nanomesh Scaffold for Ultrafast Flexible Photodetectors Based on Organic Semiconducting Crystals.
Self-Suspended Nanomesh Scaffold for Ultrafast Flexible Photodetectors Based on Organic Semiconducting Crystals.
Advanced materials (Deerfield Beach, Fla.) (2018-05-22)
Lei Zhang, Nadiia Pasthukova, Yifan Yao, Xiaolan Zhong, Egon Pavlica, Gvido Bratina, Emanuele Orgiu, Paolo Samorì
要旨
Self-standing nanostructures are of fundamental interest in materials science and nanoscience and are widely used in (opto-)electronic and photonic devices as well as in micro-electromechanical systems. To date, large-area and self-standing nanoelectrode arrays assembled on flexible substrates have not been reported. Here the fabrication of a hollow nanomesh scaffold on glass and plastic substrates with a large surface area over 1 mm2 and ultralow leakage current density (≈1-10 pA mm-2 @ 2 V) across the empty scaffold is demonstrated. Thanks to the continuous sub-micrometer space formed in between the nanomesh and the bottom electrode, highly crystalline and dendritic domains of 6,13-bis(triisopropylsilylethinyl)pentacene growing within the hollow cavity can be observed. The high degree of order at the supramolecular level leads to efficient charge and exciton transport; the photovoltaic detector supported on flexible polyethylene terephthalate substrates exhibits an ultrafast photoresponse time as short as 8 ns and a signal-to-noise ratio approaching 105 . Such a hollow scaffold holds great potential as a novel device architecture toward flexible (opto-)electronic applications based on self-assembled micro/nanocrystals.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
N,N′-ジオクチル-3,4,9,10-ペリレンジカルボキシミド, 98%