コンテンツへスキップ
Merck
  • New Flavone-Cyanoacetamide Hybrids with a Combination of Cholinergic, Antioxidant, Modulation of β-Amyloid Aggregation, and Neuroprotection Properties as Innovative Multifunctional Therapeutic Candidates for Alzheimer's Disease and Unraveling Their Mechanism of Action with Acetylcholinesterase.
New Flavone-Cyanoacetamide Hybrids with a Combination of Cholinergic, Antioxidant, Modulation of β-Amyloid Aggregation, and Neuroprotection Properties as Innovative Multifunctional Therapeutic Candidates for Alzheimer's Disease and Unraveling Their Mechanism of Action with Acetylcholinesterase.
Molecular pharmaceutics (2018-05-11)
Shaik Jeelan Basha, Penumala Mohan, Daniel Pushparaju Yeggoni, Zinka Raveendra Babu, Palaka Bhagath Kumar, Ampasala Dinakara Rao, Rajagopal Subramanyam, Amooru Gangaiah Damu
要旨
In line with the modern multi-target-directed ligand paradigm of Alzheimer's disease (AD), a series of 19 compounds composed of flavone and cyanoacetamide groups have been synthesized and evaluated as multifunctional agents against AD. Biological evaluation demonstrated that compounds 7j, 7n, 7o, 7r, and 7s exhibited excellent inhibitory potency (AChE, IC50 of 0.271 ± 0.012 to 1.006 ± 0.075 μM) and good selectivity toward acetylcholinesterase, significant antioxidant activity, good modulation effects on self-induced Aβ aggregation, low cytotoxicity, and neuroprotection in human neuroblastoma SK-N-SH cells. Further, an inclusive study on the interaction of 7j, 7n, 7o, 7r, and 7s with AChE at physiological pH 7.2 using fluorescence, circular dichroism, and molecular docking methods suggested that these derivatives bind strongly to the peripheral anionic site of AChE mostly through hydrophobic interactions. Overall, the multifunctional profiles and strong AChE binding affinity highlight these compounds as promising prototypes for further pursuit of innovative multifunctional drugs for AD.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
チオフラビンT, used as stain for amyloid
Sigma-Aldrich
フラボン, ≥99.0%