コンテンツへスキップ
Merck
  • Involvement of the bed nucleus of the stria terminalis in L-Dopa induced dyskinesia.
Involvement of the bed nucleus of the stria terminalis in L-Dopa induced dyskinesia.
Scientific reports (2017-05-26)
Matthieu F Bastide, Christelle Glangetas, Evelyne Doudnikoff, Qin Li, Mathieu Bourdenx, Pierre-Olivier Fernagut, Éric C Dumont, François Georges, Erwan Bézard
要旨
A whole brain immediate early gene mapping highlighted the dorsolateral bed nucleus of the stria terminalis (dlBST) as a structure putatively involved in L-3,4-dihydroxyphenylalanine (L-Dopa)-induced dyskinesia (LID), the debilitating side-effects of chronic dopamine replacement therapy in Parkinson's disease (PD). dlBST indeed displayed an overexpression of ∆FosB, ARC, Zif268 and FRA2 only in dyskinetic rats. We thus hypothesized that dlBST could play a role in LID hyperkinetic manifestations. To assess the causal role of the dlBST in LID, we used Daun02 inactivation to selectively inhibit the electrical activity of dlBST ΔFosB-expressing neurons. Daun02 is a prodrug converted into Daunorubicin by ß-galactosidase. Then, the newly synthesized Daunorubicin is an inhibitor of neuronal excitability. Therefore, following induction of abnormal involuntary movements (AIMs), 6-OHDA rats were injected with Daun02 in the dlBST previously expressing ß-galactosidase under control of the FosB/ΔFosB promoter. Three days after Daun02 administration, the rats were tested daily with L-Dopa to assess LID. Pharmacogenetic inactivation of ∆FosB-expressing neuron electrophysiological activity significantly reduced AIM severity. The present study highlights the role of dlBST in the rodent analog of LID, offering a new target to investigate LID pathophysiology.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
抗-D1ドーパミン受容体抗体、ラットモノクローナル ラット宿主抗体, clone 1-1-F11 s.E6, purified from hybridoma cell culture