Merck
22941
4-Port Sampling and Switching Valve
1/16 in. fitting, maximum temperature 300 °C
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41112500
eCl@ss:
32060101
物質
(Nitronic 60)
特徴
number of ports 4
メーカー/製品名
C4T
VICI Valco®
最高温度
300 °C
フィッティング
1/16 in. fitting
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Sampling valve basically supports the discrete volumes of the treated samples stream to periodically inject into the carrier gas stream of the chromatograph. Sampling valves are fitted with electric or pneumatic actuators which help them to be operated automatically. Switching valve are critical if binary pumps are used. It allows employment of different desired mobile phases. Switching valve consists of five parts: control unit, connecting device, valve body, a rotor and preloaded assembly.
カラムを隔離して、上流のカラムから出た化合物が不可逆的に吸着するのを防ぎます。図では、CO2がカラム1内の空気から分離され、弁が位置Bにあればカラム2を迂回します。CO2が検出されると、弁が位置Aに戻り、O2、N2、CH4、およびCOがカラム2から検出器に溶出します。
アプリケーション
Sampling valve may be used to connect the LC and GC instruments to allow fast and dependable introduction of large sample volumes onto a capillary GC column.
法的情報
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
試験成績書(COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact カスタマーサポート
以前この製品を購入いただいたことがある場合
文書ライブラリで、最近購入した製品の文書を検索できます。
文書ライブラリにアクセスする
Clément Vinauger et al.
Current biology : CB, 28(3), 333-344 (2018-02-06)
How mosquitoes determine which individuals to bite has important epidemiological consequences. This choice is not random; most mosquitoes specialize in one or a few vertebrate host species, and some individuals in a host population are preferred over others. Mosquitoes will
On-line combination of liquid chromatography and capillary gas chromatography. Preconcentration and analysis of organic compounds in aqueous samples.
Noy, Th, et al.
Journal of High Resolution Chromatography, 11, 181-186 (1988)
Walt Boyes
Instrumentation Reference Book, 414-414 (2009)
Satinder Ahuja
Chiral Separation Methods for Pharmaceutical and Biotechnological Products, 218-218 (2011)