Merck
Z248762
電卓、Hewlett-Packard
Model HP 48GX Expandable Graphic Calculator
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
44101800
メーカー/製品名
Texas Instruments
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
A selection of hand-held calculators with features for a wide variety of scientific applications. Hundreds of pre-programmed functions plus user programmability give the power and versatility of a computer in a calculator-sized package.
Except as noted, all HP calculators below have these features:
  • User programmable, with prompts, conditional branches, indexed loops, subroutines
  • RPN entry logic
  • Functions: math, exponential, trig, inverse trig, hyperbolics, numerical integration (area under a curve), HP solve (root-finder), complex number operations, angle/time conversions, base conversions, statistical functions (standard deviation, linear regression); combinations/permutations
All features are as for the HP-48G above, but with additional capabilities through plug-in expansion options: 128K RAM expandable to 4 MB.
法的情報
Hewlett-Packard is a trademark of Hewlett-Packard Corp.
最新バージョンのいずれかを選択してください:
試験成績書(COA)