Merck
GEID2212101
GE Ettan DALT gel cassette
gel size 25 cm × 20 cm
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41105300
NACRES:
NB.22
包装
pack of 1 ea
メーカー/製品名
Cytiva Article No. 80-6067-27
Cytiva ID2212-1,0-1
ゲルサイズ
25 cm × 20 cm
詳細
Rquires no assembly, the two glass plates are attached along one edge by a flexible silicone rubber hinge and the spacers, 1.0- or 1.5-mm thick, are permanently attached to one of the glass plates. No clamps or tape are required in the DALT multi-gel casting unit.
法的情報
Ettan is a trademark of Cytiva
最新バージョンのいずれかを選択してください:
試験成績書(COA)