Merck
D1045
Sigma-Aldrich
Pipette tip disposal container, acrylic
inside L 51/4 in. 13.1 cm W 6 in. 15.2 cm H 61/8 in. 15.5 cm
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
26142300
詳細
Made of 3/8 in.acrylic for shielded disposal of β-emitter contaminated pipet tips. A hinged gate covers the small port. Eject the contaminated tip from the pipetter then close the gate to ensure complete shielding.
最新バージョンのいずれかを選択してください:
試験成績書(COA)