Merck
CLS10044
Corning® HYPERStack®スタッキングトレー、オレンジ、ABS材
no adhesives, low particulate
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41100000
NACRES:
NA.75
物質
(Acrylonitrile Butadiene Styrene, ABS)
メーカー/製品名
Corning 10044
詳細
Features and Benefits:
More cells – 2.5x more cells per volumetric footprint
Closed system – No open fluid manipulations
Scalable product – Multiple size offerings
Ergonomic design – Easier manipulation
Fixed media volume – 0.2 mL/cm2 fills vessel for optimal handling
Innovative assembly – No adhesives, low particulate
Lower disposal cost – Less volumetric waste per growth area
法的情報
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)