包装
case of 5 packs
pkg of 960 tips (racked)
容積測定範囲
0.1-10 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Art 10 Reachはチップが長く、サンプル・チュ-ブ・ピペットのコンタミネ-ションを防ぎます。
包装
960EA = 96チップが10トレイ(1パック)
4800EA = 960チップが5パック(96チップが50トレイ)
800EA = 100チップが8トレイ(1パック)
3200EA = 800EAが4パック
4800EA = 960チップが5パック(96チップが50トレイ)
800EA = 100チップが8トレイ(1パック)
3200EA = 800EAが4パック
適合性
適合性
Biohit mLINE 0.5-10 μL
Biohit Electronic 0.2-10 μL
Biohit Variable Single/Multi-Channel 0.5-10 μL
Capp Aero 10 μL
Corning 4959 0.1-2 μL
Corning 4960 0.5-10 μL
Eppendorf Reference/Research 0.1-2.5 μL
Finnpipette 4500 Digital 0.2-2 μL
Finnpipette 4500 Digital Micro 0.5-10 μL
Finnpipette Focus 4600 3uL, 5uL, 10 μL
Finnpipette Multi-Channel 0.5-10 μL
Hamilton SoftGrip Precision wul/10 μL
Mettler-Toledo VoluMate 0.1-2.5 μL
Nichiryo BenchMate 0.2-2 μL/0.5-10 μL
Pipetman P-2 0.1-2 μL
Pipetman P-10 0.5-10 μL
Rainin EDP 2 0.5-10 μL
Rainin EDP Plus 0.5 -10 μL
Rainin Pipet Plus 0.2-2 μL/0.5-10 μL
Rainin Pipetman 0.2-2 μL/0.5-10 μL
Transferpette Single/Multi-Channel 0.5-10 μL/2-20 μL
Transferpette Ultra-Micro 2-20 μL
Biohit mLINE 0.5-10 μL
Biohit Electronic 0.2-10 μL
Biohit Variable Single/Multi-Channel 0.5-10 μL
Capp Aero 10 μL
Corning 4959 0.1-2 μL
Corning 4960 0.5-10 μL
Eppendorf Reference/Research 0.1-2.5 μL
Finnpipette 4500 Digital 0.2-2 μL
Finnpipette 4500 Digital Micro 0.5-10 μL
Finnpipette Focus 4600 3uL, 5uL, 10 μL
Finnpipette Multi-Channel 0.5-10 μL
Hamilton SoftGrip Precision wul/10 μL
Mettler-Toledo VoluMate 0.1-2.5 μL
Nichiryo BenchMate 0.2-2 μL/0.5-10 μL
Pipetman P-2 0.1-2 μL
Pipetman P-10 0.5-10 μL
Rainin EDP 2 0.5-10 μL
Rainin EDP Plus 0.5 -10 μL
Rainin Pipet Plus 0.2-2 μL/0.5-10 μL
Rainin Pipetman 0.2-2 μL/0.5-10 μL
Transferpette Single/Multi-Channel 0.5-10 μL/2-20 μL
Transferpette Ultra-Micro 2-20 μL
法的情報
ART is a registered trademark of Molecular BioProducts, Inc.