Merck
MSLM25001
Millipore
MilliSnap® System Flat White Adapter
for Incubation on Liquid Media Cassette
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41116129
NACRES:
NB.24
品質水準
メーカー/製品名
MilliSnap®
適合性
for use with MilliSnap® System
詳細
MilliSnap® System Flat White Adapter is an accessory component designed to simplify workflow intended while performing bioburden testing, water monitoring, pharmaceutical product testing etc. The white adapter belongs to a plug and play hands free system which will separate Milliflex® funnels and attach the filtered membrane onto media cassettes. The white adapter also ensures complete contact between the membrane and media for enhanced results.
アプリケーション
MilliSnap® System Flat White Adapter is used to separate filtered Milliflex® membranes onto media cassettes in a hands-free manner while performing bioburden testing, water monitoring, pharmaceutical product testing etc.
法的情報
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLISNAP is a registered trademark of Merck KGaA, Darmstadt, Germany