この製品は、Merck Milliporeウェブサイトでのみ注文できます。
製品名
Knurled nuts with long bushing and PVDF cones for 6 port valves (Rheodyne),
technique(s)
HPLC: suitable
Quality Segment
storage temp.
no temp limit
General description
The package contains: 2 knurled nuts; 6 PVDF double cones.
Analysis Note
visual control: passes test
1 of 1
当該品目 | |||
|---|---|---|---|
| technique(s) HPLC: suitable | technique(s) HPLC: suitable | technique(s) HPLC: suitable | technique(s) HPLC: suitable |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. no temp limit | storage temp. no temp limit | storage temp. no temp limit | storage temp. no temp limit |
Still not finding the right product?
Explore all of our products under Knurled nuts with long bushing and PVDF cones for 6 port valves (Rheodyne)