Merck
Z167606
Cover opener for plastic pails
aluminum (cast)
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121800
NACRES:
NB.51
物質
aluminum (cast)
詳細
Durable cast aluminum tool (W x L 1 1/2 in. x 8 1/4 in.) grabs cover tightly for opening. Leaves cover intact for re-closing. Integral finger grooves on the handle provide a non-slip grip. Small enough to carry in a pocket.
最新バージョンのいずれかを選択してください:
試験成績書(COA)