HPLC Analysis of Serine Enantiomers on Astec® CHIROBIOTIC® TAG
![HPLC Analysis of Serine Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC](/deepweb/assets/sigmaaldrich/product/images/150/857/e8b43584-8ae4-41a8-87ac-2e5fcf0f8b57/800/e8b43584-8ae4-41a8-87ac-2e5fcf0f8b57.jpg)
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] water; [B] acetonitrile; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
5 μL
sample
10 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC