Merck
  • Transition metal salts-catalyzed aza-Michael reactions of enones with carbamates.
Transition metal salts-catalyzed aza-Michael reactions of enones with carbamates.
Organic letters (2002-04-13)
Shū Kobayashi, Kentaro Kakumoto, Masaharu Sugiura
ABSTRACT
Several transition metal salts were found to catalyze aza-Michael reactions of enones with carbamates efficiently. The catalytic activity was strongly dependent on the nature of the metal salts. While conventional Lewis acids such as BF(3).OEt(2), AlCl(3), or TiCl(4) showed lower activity, group 7-11 transition metal salts in higher oxidation states such as ReCl(5), Fe(ClO(4))(3).9H(2)O, RuCl(3).nH(2)O, OsCl(3).3H(2)O, RhCl(3).nH(2)O, PtCl(4).5H(2)O, or AuCl(3).H(2)O exhibited higher catalytic activity. [reaction: see text]
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%
Pricing and availability is not currently available.