Merck
  • Understanding the Adsorption of PFOA on MIL-101(Cr)-Based Anionic-Exchange Metal-Organic Frameworks: Comparing DFT Calculations with Aqueous Sorption Experiments.
Understanding the Adsorption of PFOA on MIL-101(Cr)-Based Anionic-Exchange Metal-Organic Frameworks: Comparing DFT Calculations with Aqueous Sorption Experiments.
Environmental science & technology (2015-06-13)
Kai Liu, Siyu Zhang, Xiyue Hu, Kunyang Zhang, Ajay Roy, Gang Yu
ABSTRACT
To examine the effects of different functionalization methods on adsorption behavior, anionic-exchange MIL-101(Cr) metal-organic frameworks (MOFs) were synthesized using preassembled modification (PAM) and postsynthetic modification (PSM) methods. Perfluorooctanoic acid (PFOA) adsorption results indicated that the maximum PFOA adsorption capacity was 1.19 and 1.89 mmol g(-1) for anionic-exchange MIL-101(Cr) prepared by PAM and PSM, respectively. The sorption equilibrium was rapidly reached within 60 min. Our results indicated that PSM is a better modification technique for introducing functional groups onto MOFs for adsorptive removal because PAM places functional groups onto the aperture of the nanopore, which hinders the entrance of organic contaminants. Our experimental results and the results of complementary density functional theory calculations revealed that in addition to the anion-exchange mechanism, the major PFOA adsorption mechanism is a combination of Lewis acid/base complexation between PFOA and Cr(III) and electrostatic interaction between PFOA and the protonated carboxyl groups of the bdc (terephthalic acid) linker.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, SAJ special grade
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol, NMR reference standard
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20