Merck
  • Development and In Vitro Evaluation of Vitamin E-Enriched Nanoemulsion Vehicles Loaded with Genistein for Chemoprevention Against UVB-Induced Skin Damage.
Development and In Vitro Evaluation of Vitamin E-Enriched Nanoemulsion Vehicles Loaded with Genistein for Chemoprevention Against UVB-Induced Skin Damage.
Journal of pharmaceutical sciences (2015-06-26)
Bill Brownlow, Vinay J Nagaraj, Amy Nayel, Megha Joshi, Tamer Elbayoumi
ABSTRACT
There is a great need for effective protection against cutaneous pathologies arising from chronic exposure to harmful solar UVB radiations. A promising pharmaceutical strategy to improve the efficacy of chemotherapeutic/preventative natural compounds (e.g., soy isoflavone Genistein, Gen) is to enhance their dermal delivery using nanoemulsion (NE) formulations. This report investigates the development of nanoemulsified tocotrienol(T3)-rich fraction of red palm oil (Tocomin®), to yield an optimal NE delivery system for dermal photoprotection (z-average size <150 nm, ζ-potential ≈ -30 mV, polydispersity index < 0.25). Physicochemical characterization and photostability studies indicate NE formulations utilizing surfactant mixture (Smix) of Solutol® HS-15 (SHS15) blended with vitamin E TPGS (TPGS) as cosurfactant was significantly superior to formulations that utilized Lutrol® F68 (LF68) as the cosurfactant. A ratio of 60:40 of SHS15-TPGS-NE was further identified as lead Tocomin® NE topical platform using in vitro pharmaceutical skin reactivity studies that assess cutaneous irritancy and cytotoxicity. Prototype Tocomin® NE loaded with the antiphotocarcinogenic molecule Gen (Gen-Tocomin® NE) showed slow-release profile in both liquid and cream forms. Gen-Tocomin® NE also showed excellent biocompatibility, and provided substantial UVB protection to cultured subcutaneous L929 fibroblasts, indicating the great potential of our Tocomin® NE warranting further prototype development as topical pharmaceutical platform for skin photoprotection applications.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%