Merck
  • CD24+ Ovarian Cancer Cells Are Enriched for Cancer-Initiating Cells and Dependent on JAK2 Signaling for Growth and Metastasis.
CD24+ Ovarian Cancer Cells Are Enriched for Cancer-Initiating Cells and Dependent on JAK2 Signaling for Growth and Metastasis.
Molecular cancer therapeutics (2015-05-15)
Daniela Burgos-Ojeda, Rong Wu, Karen McLean, Yu-Chih Chen, Moshe Talpaz, Euisik Yoon, Kathleen R Cho, Ronald J Buckanovich
ABSTRACT
Ovarian cancer is known to be composed of distinct populations of cancer cells, some of which demonstrate increased capacity for cancer initiation and/or metastasis. The study of human cancer cell populations is difficult due to long requirements for tumor growth, interpatient variability, and the need for tumor growth in immune-deficient mice. We therefore characterized the cancer initiation capacity of distinct cancer cell populations in a transgenic murine model of ovarian cancer. In this model, conditional deletion of Apc, Pten, and Trp53 in the ovarian surface epithelium (OSE) results in the generation of high-grade metastatic ovarian carcinomas. Cell lines derived from these murine tumors express numerous putative stem cell markers, including CD24, CD44, CD90, CD117, CD133, and ALDH. We show that CD24(+) and CD133(+) cells have increased tumor sphere-forming capacity. CD133(+) cells demonstrated a trend for increased tumor initiation while CD24(+) cells versus CD24(-) cells had significantly greater tumor initiation and tumor growth capacity. No preferential tumor-initiating or growth capacity was observed for CD44(+), CD90(+), CD117(+), or ALDH(+) versus their negative counterparts. We have found that CD24(+) cells, compared with CD24(-) cells, have increased phosphorylation of STAT3 and increased expression of STAT3 target Nanog and c-myc. JAK2 inhibition of STAT3 phosphorylation preferentially induced cytotoxicity in CD24(+) cells. In vivo JAK2 inhibitor therapy dramatically reduced tumor metastases, and prolonged overall survival. These findings indicate that CD24(+) cells play a role in tumor migration and metastasis and support JAK2 as a therapeutic target in ovarian cancer.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Luciferin, synthetic
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DAPI, for nucleic acid staining
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)