Merck
  • HPLC Method for Simultaneous Quantification of Bakuchiol and Minor Furocoumarins in Bakuchiol Extract from Psoralea corylifolia.
HPLC Method for Simultaneous Quantification of Bakuchiol and Minor Furocoumarins in Bakuchiol Extract from Psoralea corylifolia.
Journal of AOAC International (2015-08-14)
Mijeong Jeong, Thao Hong, Kangwoo Lee, Hyunjung Hwangbo, Minjung Kim, Wenwen Ma, Michael Zahn
ABSTRACT
A simple, sensitive, and rapid HPLC method was developed to analyze bakuchiol and two furocoumarins (psoralen and angelicin) simultaneously in bakuchiol extracts from Psoralea corylifolia seeds. The analysis was performed within 30 min on a phenyl-hexyl column using gradient elution with a mobile phase composed of water and methanol with UV detection at 260 nm for bakuchiol and 246 nm for psoralen and angelicin. The method was validated with respect to linearity (r2>0.99 for all components), accuracy (>95% for all components), and precision (<2% RSD for both interday and intraday). Sensitivity of impurity detection in the sample was achieved as low as 0.36 and 0.31 μg/mL for psoralen and angelicin, respectively. Therefore, the method is suitable for QC of P. corylifolia extracts and bakuchiol related samples.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%