Merck
  • Bisphenol A contamination in soft drinks as a risk for children's health in Italy.
Bisphenol A contamination in soft drinks as a risk for children's health in Italy.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-04-24)
Evelina Fasano, Francesco Esposito, Gelsomina Scognamiglio, Fabio Di Francesco, Paolo Montuori, Renata Amodio Cocchieri, Teresa Cirillo
ABSTRACT
Bisphenol A (BPA) was determined in sugary carbonated, non-carbonated and milk-based beverages, through HLPC-fluorescence detection and confirmed by LC-MS/MS, in a selection of brands that are mostly consumed by Italian children. The daily intake was determined through the WHO budget method (BM). BPA was found at detectable levels in 57% of carbonated beverages, in 50% of non-carbonated and in 100% of milk-based beverages. The median concentrations were 1.24 µg l(-1) (range = < LOD-4.98 µg l(-1)) in canned carbonated beverages and 0.18 µg l(-1) (< LOD-1.78 µg l(-1)) in non-canned carbonated beverages. In non-carbonated beverages, median concentrations were 0.80 µg l(-1) (< LOD-2.79 µg l(-1)) and 0.18 µg l(-1) (< LOD-3.58 µg l(-1)), respectively, for canned and non-canned beverages; in milk-based products the BPA median concentration was 3.60 µg l(-1) (1.00-17.65 µg l(-1)). BPA daily intake from sugary drink consumption in children ranged from 0.008 to 1.765 µg kg(-1) bw day(-1). The median exposure values for the 'best' and 'worst' cases were 0.16% and 0.47% respectively of the EFSA t-TDI for BPA (4 µg kg(-1) bw day(-1)), and 10.59% and 35.30% of the t-TDI when the maximum levels were considered.
Sigma-Aldrich
Octadecylsilane, 97%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, NMR reference standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Pricing and availability is not currently available. Contact Technical Service
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available. Contact Technical Service