Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Rhodamine B, for fluorescence
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroacetic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Pricing and availability is not currently available.
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium permanganate, suitable for determination of toxic metals, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, suitable for determination of toxic metals, ≥35.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, JIS special grade, 30.0-35.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, SAJ first grade, 32.2-36.8% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 3% in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.0125 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium permanganate, SAJ first grade, ≥99.3%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, SAJ first grade, 9.5-10.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium nitrate, JIS special grade, ≥99.0%
Pricing and availability is not currently available.